Assay Details
| Gene Symbol: TAFII15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342109.1 | NP_001329548.1 | 3 - 4 | 538 | 98 | |
| NM_119321.4 | NP_194900.1 | 3 - 4 | 429 | 98 | ||
| NM_179150.4 | NP_849481.1 | 3 - 4 | 412 | 98 | ||
| GenBank mRNA | AF344879.1 | - | 3 - 4 | 339 | 98 | |
| AF352813.1 | - | 3 - 4 | 339 | 98 | ||
| AK317292.1 | - | 3 - 4 | 344 | 98 | ||
| AY086560.1 | - | 3 - 4 | 344 | 98 | ||
| AY463628.1 | - | 2 - 3 | 152 | 98 | ||
| Y12904.1 | - | 329 | 98 | |||
More Information
Set Membership: |
|

TAFII15
Genomic Map