Assay Details
| Gene Symbol: TAFII15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342109.1 | NP_001329548.1 | 5 - 6 | 661 | 94 | |
| NM_119321.4 | NP_194900.1 | 5 - 6 | 552 | 94 | ||
| NM_179150.4 | NP_849481.1 | 5 - 6 | 535 | 94 | ||
| GenBank mRNA | AF344879.1 | - | 5 - 6 | 462 | 94 | |
| AF352813.1 | - | 5 - 6 | 462 | 94 | ||
| AK317292.1 | - | 5 - 6 | 467 | 94 | ||
| AY086560.1 | - | 5 - 6 | 467 | 94 | ||
| AY463628.1 | - | 4 - 5 | 275 | 94 | ||
| Y12904.1 | - | 452 | 94 | |||
More Information
Set Membership: |
|

TAFII15
Genomic Map