Assay Details
| Gene Symbol: AT4G33150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342200.1 | NP_001328161.1 | 12 - 13 | 1722 | 164 | |
| NM_119469.4 | NP_567914.1 | 13 - 14 | 1680 | 164 | ||
| NM_179155.4 | NP_849486.1 | 13 - 14 | 1801 | 164 | ||
More Information
Set Membership: |
|

AT4G33150
Genomic Map