Assay Details
| Gene Symbol: EX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342225.1 | NP_001328575.1 | 4 - 5 | 811 | 65 | |
| NM_119519.4 | NP_567929.1 | 4 - 5 | 961 | 65 | ||
| NM_179157.3 | NP_849488.1 | 4 - 5 | 961 | 65 | ||
| GenBank mRNA | AY059742.1 | - | 4 - 5 | 750 | 65 | |
| AY117173.1 | - | 4 - 5 | 693 | 65 | ||
More Information
Set Membership: |
|

EX1
Genomic Map