Assay Details
| Gene Symbol: AGB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119611.5 | NP_195172.1 | 1 - 2 | 456 | 72 | |
| NM_179163.3 | NP_849494.2 | 1 - 2 | 340 | 72 | ||
| NM_202954.3 | NP_974683.1 | 1 - 2 | 342 | 72 | ||
| GenBank mRNA | U12232.1 | - | 1 - 2 | 309 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_179163.3 transcripts.
Set Membership: |
|

AGB1
Genomic Map