Assay Details
| Gene Symbol: RecQl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119740.3 | NP_195299.2 | 12 - 13 | 1355 | 150 | |
| NM_179169.1 | NP_849500.1 | 11 - 12 | 1215 | 150 | ||
| GenBank mRNA | AJ404472.1 | - | 12 - 13 | 1223 | 150 | |
More Information
Set Membership: |
|

RecQl3
Genomic Map