Assay Details
| Gene Symbol: RecQl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119740.3 | NP_195299.2 | 14 - 15 | 1494 | 125 | |
| NM_179169.1 | NP_849500.1 | 13 - 14 | 1354 | 125 | ||
| GenBank mRNA | AJ404472.1 | - | 14 - 15 | 1362 | 125 | |
More Information
Set Membership: |
|

RecQl3
Genomic Map