Assay Details
| Gene Symbol: BGAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119799.4 | NP_568001.1 | 10 - 11 | 1424 | 85 | |
| NM_179175.2 | NP_849506.1 | 10 - 11 | 1219 | 85 | ||
| GenBank mRNA | AJ270299.1 | - | 10 - 11 | 1141 | 85 | |
| AY056285.1 | - | 10 - 11 | 1200 | 85 | ||
| AY091432.1 | - | 10 - 11 | 1101 | 85 | ||
More Information
Set Membership: |
|

BGAL3
Genomic Map