Assay Details
| Gene Symbol: BGAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119799.4 | NP_568001.1 | 12 - 13 | 1769 | 71 | |
| NM_179175.2 | NP_849506.1 | 12 - 13 | 1564 | 71 | ||
| GenBank mRNA | AJ270299.1 | - | 12 - 13 | 1486 | 71 | |
| AY056285.1 | - | 12 - 13 | 1545 | 71 | ||
| AY091432.1 | - | 12 - 13 | 1446 | 71 | ||
More Information
Set Membership: |
|

BGAL3
Genomic Map