Assay Details
| Gene Symbol: BGAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119799.4 | NP_568001.1 | 15 - 16 | 2100 | 85 | |
| NM_179175.2 | NP_849506.1 | 15 - 16 | 1895 | 85 | ||
| GenBank mRNA | AJ270299.1 | - | 15 - 16 | 1817 | 85 | |
| AY056285.1 | - | 15 - 16 | 1876 | 85 | ||
| AY091432.1 | - | 15 - 16 | 1777 | 85 | ||
More Information
Set Membership: |
|

BGAL3
Genomic Map