Assay Details
| Gene Symbol: BGAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119799.4 | NP_568001.1 | 16 - 17 | 2208 | 73 | |
| NM_179175.2 | NP_849506.1 | 16 - 17 | 2003 | 73 | ||
| GenBank mRNA | AJ270299.1 | - | 16 - 17 | 1925 | 73 | |
| AY056285.1 | - | 16 - 17 | 1984 | 73 | ||
| AY091432.1 | - | 16 - 17 | 1885 | 73 | ||
More Information
Set Membership: |
|

BGAL3
Genomic Map