Assay Details
| Gene Symbol: nPAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036701.1 | NP_001031778.1 | 8 - 9 | 1410 | 75 | |
| NM_001036702.1 | NP_001031779.1 | 8 - 9 | 1418 | 75 | ||
| NM_001125630.1 | NP_001119102.1 | 8 - 9 | 1410 | 75 | ||
| NM_001125631.2 | NP_001119103.1 | 8 - 9 | 1479 | 75 | ||
| NM_001342179.1 | NP_001329665.1 | 8 - 9 | 1479 | 75 | ||
| NM_001342180.1 | NP_001329664.1 | 8 - 9 | 1479 | 75 | ||
| NM_001342181.1 | NP_001329663.1 | 8 - 9 | 1479 | 75 | ||
| NM_001342182.1 | NP_001329669.1 | 8 - 9 | 1479 | 75 | ||
| NM_001342183.1 | NP_001329668.1 | 8 - 9 | 1469 | 75 | ||
| NM_001342184.1 | NP_001329667.1 | 8 - 9 | 1469 | 75 | ||
| NM_001342185.1 | NP_001329666.1 | 8 - 9 | 1445 | 75 | ||
| NM_119438.4 | NP_195011.4 | 8 - 9 | 1410 | 75 | ||
| NM_179230.2 | NP_849561.1 | 8 - 9 | 1479 | 75 | ||
| NM_179231.2 | NP_849562.2 | 8 - 9 | 1410 | 75 | ||
| NM_202939.3 | NP_974668.1 | 8 - 9 | 1479 | 75 | ||
| NM_202940.3 | NP_974669.1 | 8 - 9 | 1479 | 75 | ||
| NM_202941.3 | NP_974670.2 | 8 - 9 | 1479 | 75 | ||
| GenBank mRNA | AK316738.1 | - | 8 - 9 | 1411 | 75 | |
| AK318704.1 | - | 8 - 9 | 1415 | 75 | ||
| AY069880.1 | - | 8 - 9 | 1410 | 75 | ||
| BT000485.1 | - | 8 - 9 | 1056 | 75 | ||
More Information
Set Membership: |
|

nPAP
Genomic Map