Assay Details
| Gene Symbol: nPAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125630.1 | NP_001119102.1 | 13 - 14 | 2678 | 77 | |
| NM_119438.4 | NP_195011.4 | 13 - 14 | 2648 | 77 | ||
| NM_179231.2 | NP_849562.2 | 13 - 14 | 2657 | 77 | ||
More Information
Set Membership: |
|

nPAP
Genomic Map