Assay Details
| Gene Symbol: CHB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119608.3 | NP_195169.3 | 5 - 6 | 2558 | 115 | |
| NM_179232.1 | NP_849563.1 | 5 - 6 | 2566 | 115 | ||
| NM_179233.2 | NP_849564.1 | 5 - 6 | 2579 | 115 | ||
| NM_202953.1 | NP_974682.1 | 5 - 6 | 2566 | 115 | ||
| GenBank mRNA | AY074306.1 | - | 5 - 6 | 2566 | 115 | |
| AY096454.1 | - | 5 - 6 | 2417 | 115 | ||
More Information
Set Membership: |
|

CHB3
Genomic Map