Assay Details
| Gene Symbol: BGLU11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179247.6 | NP_849578.5 | 7 - 8 | 943 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BGLU11| Gene Symbol: BGLU11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179247.6 | NP_849578.5 | 7 - 8 | 943 | 94 | |
Set Membership: |
|