Assay Details
| Gene Symbol: PRMT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100365.4 | NP_563720.1 | 3 - 4 | 737 | 63 | |
| NM_179260.1 | NP_849591.1 | 3 - 4 | 648 | 63 | ||
| GenBank mRNA | AY058846.1 | - | 3 - 4 | 617 | 63 | |
| AY132011.1 | - | 3 - 4 | 514 | 63 | ||
More Information
Set Membership: |
|

PRMT10
Genomic Map