Assay Details
| Gene Symbol: TAFII59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035892.3 | NP_001030969.1 | 11 - 12 | 2007 | 117 | |
| NM_100373.4 | NP_171987.1 | 12 - 13 | 1899 | 117 | ||
| NM_179261.4 | NP_849592.1 | 12 - 13 | 1785 | 117 | ||
| GenBank mRNA | AY050962.1 | - | 12 - 13 | 1828 | 117 | |
More Information
Set Membership: |
|

TAFII59
Genomic Map