Assay Details
| Gene Symbol: TAFII59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035892.3 | NP_001030969.1 | 3 - 4 | 667 | 105 | |
| NM_100373.4 | NP_171987.1 | 4 - 5 | 559 | 105 | ||
| NM_179261.4 | NP_849592.1 | 4 - 5 | 445 | 105 | ||
| GenBank mRNA | AY050962.1 | - | 4 - 5 | 488 | 105 | |
| AY091292.1 | - | 3 - 4 | 321 | 105 | ||
| AY463621.1 | - | 3 - 4 | 321 | 105 | ||
More Information
Set Membership: |
|

TAFII59
Genomic Map