Assay Details
| Gene Symbol: SR45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331692.1 | NP_001323001.1 | 2 - 3 | 188 | 65 | |
| NM_001331693.1 | NP_001323003.1 | 2 - 3 | 277 | 65 | ||
| NM_001331694.1 | NP_001323002.1 | 2 - 3 | 277 | 65 | ||
| NM_001331695.1 | NP_001323004.1 | 2 - 3 | 179 | 65 | ||
| NM_100609.4 | NP_563787.2 | 2 - 3 | 277 | 65 | ||
| NM_179274.2 | NP_849605.1 | 2 - 3 | 188 | 65 | ||
| GenBank mRNA | AY053418.1 | - | 2 - 3 | 179 | 65 | |
| BT005797.1 | - | 2 - 3 | 191 | 65 | ||
More Information
Set Membership: |
|

SR45a
Genomic Map