Assay Details
| Gene Symbol: SR45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179274.2 | NP_849605.1 | 5 - 6 | 543 | 72 | |
| GenBank mRNA | AY053418.1 | - | 5 - 6 | 534 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SR45a| Gene Symbol: SR45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179274.2 | NP_849605.1 | 5 - 6 | 543 | 72 | |
| GenBank mRNA | AY053418.1 | - | 5 - 6 | 534 | 72 | |
Set Membership: |
|