Assay Details
| Gene Symbol: SR45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331692.1 | NP_001323001.1 | 5 - 6 | 748 | 104 | |
| NM_001331693.1 | NP_001323003.1 | 4 - 5 | 994 | 104 | ||
| NM_001331694.1 | NP_001323002.1 | 5 - 6 | 837 | 104 | ||
| NM_001331695.1 | NP_001323004.1 | 4 - 5 | 896 | 104 | ||
| NM_100609.4 | NP_563787.2 | 5 - 6 | 594 | 104 | ||
| NM_179274.2 | NP_849605.1 | 6 - 7 | 601 | 104 | ||
| GenBank mRNA | AY053418.1 | - | 6 - 7 | 592 | 104 | |
| BT005797.1 | - | 5 - 6 | 508 | 104 | ||
More Information
Set Membership: |
|

SR45a
Genomic Map