Assay Details
| Gene Symbol: AT1G08315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179282.2 | NP_849613.1 | 1 | 762 | 88 | |
| GenBank mRNA | AK229288.1 | - | 1 | 715 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G08315| Gene Symbol: AT1G08315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179282.2 | NP_849613.1 | 1 | 762 | 88 | |
| GenBank mRNA | AK229288.1 | - | 1 | 715 | 88 | |
Set Membership: |
|