Assay Details
| Gene Symbol: THA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035924.2 | NP_001031001.2 | 3 - 4 | 569 | 86 | |
| NM_001084023.1 | NP_001077492.1 | 3 - 4 | 609 | 86 | ||
| NM_001331780.1 | NP_001318956.1 | 2 - 3 | 580 | 86 | ||
| NM_001331781.1 | NP_001322023.1 | 3 - 4 | 530 | 86 | ||
| NM_100736.3 | NP_563822.1 | 3 - 4 | 1046 | 86 | ||
| NM_179283.2 | NP_849614.1 | 3 - 4 | 609 | 86 | ||
| NM_179284.4 | NP_849615.1 | 3 - 4 | 642 | 86 | ||
| GenBank mRNA | AK230078.1 | - | 3 - 4 | 651 | 86 | |
| AK317034.1 | - | 3 - 4 | 574 | 86 | ||
| AY080670.1 | - | 3 - 4 | 605 | 86 | ||
| AY117227.1 | - | 1 - 2 | 440 | 86 | ||
More Information
Set Membership: |
|

THA1
Genomic Map