Assay Details
| Gene Symbol: THA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035924.2 | NP_001031001.2 | 6 - 7 | 935 | 87 | |
| NM_001331780.1 | NP_001318956.1 | 5 - 6 | 946 | 87 | ||
| NM_001331781.1 | NP_001322023.1 | 6 - 7 | 896 | 87 | ||
| NM_100736.3 | NP_563822.1 | 6 - 7 | 1412 | 87 | ||
| NM_179283.2 | NP_849614.1 | 6 - 7 | 975 | 87 | ||
| NM_179284.4 | NP_849615.1 | 6 - 7 | 1008 | 87 | ||
| GenBank mRNA | AK230078.1 | - | 6 - 7 | 1017 | 87 | |
| AY080670.1 | - | 6 - 7 | 971 | 87 | ||
| AY117227.1 | - | 4 - 5 | 806 | 87 | ||
More Information
Set Membership: |
|

THA1
Genomic Map