Assay Details
| Gene Symbol: PIF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331836.1 | NP_001318964.1 | 6 - 7 | 1547 | 77 | |
| NM_001331837.1 | NP_001318965.1 | 6 - 7 | 1607 | 77 | ||
| NM_001331838.1 | NP_001322700.1 | 5 - 6 | 1971 | 77 | ||
| NM_001331839.1 | NP_001322701.1 | 4 - 5 | 1944 | 77 | ||
| NM_100824.3 | NP_172424.1 | 5 - 6 | 1520 | 77 | ||
| NM_179295.3 | NP_849626.1 | 5 - 6 | 1580 | 77 | ||
| GenBank mRNA | AF100166.1 | - | 5 - 6 | 1212 | 77 | |
| AF251693.1 | - | 4 - 5 | 1129 | 77 | ||
| AK117255.1 | - | 5 - 6 | 1265 | 77 | ||
More Information
Set Membership: |
|

PIF3
Genomic Map