Assay Details
| Gene Symbol: BCAT-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123785.1 | NP_001117257.1 | 6 - 7 | 1416 | 119 | |
| NM_001331880.1 | NP_001320930.1 | 8 - 9 | 1290 | 119 | ||
| NM_001331881.1 | NP_001320929.1 | 8 - 9 | 1290 | 119 | ||
| NM_001331883.1 | NP_001320927.1 | 8 - 9 | 1124 | 119 | ||
| NM_179298.2 | NP_849629.1 | 8 - 9 | 1124 | 119 | ||
| GenBank mRNA | AJ276123.1 | - | 8 - 9 | 1004 | 119 | |
| AK316768.1 | - | 8 - 9 | 1053 | 119 | ||
More Information
Set Membership: |
|

BCAT-1
Genomic Map