Assay Details
| Gene Symbol: BCAT-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123785.1 | NP_001117257.1 | 7 - 8 | 1539 | 81 | |
| NM_001331881.1 | NP_001320929.1 | 9 - 10 | 1413 | 81 | ||
| NM_001331882.1 | NP_001320928.1 | 9 - 10 | 1335 | 81 | ||
| NM_100880.3 | NP_563859.1 | 9 - 10 | 1206 | 81 | ||
| NM_179298.2 | NP_849629.1 | 9 - 10 | 1247 | 81 | ||
| GenBank mRNA | AJ276123.1 | - | 9 - 10 | 1127 | 81 | |
| AK316768.1 | - | 9 - 10 | 1176 | 81 | ||
| AY054517.1 | - | 9 - 10 | 1207 | 81 | ||
| AY128778.1 | - | 9 - 10 | 1000 | 81 | ||
More Information
Set Membership: |
|

BCAT-1
Genomic Map