Assay Details
| Gene Symbol: PHR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101109.1 | NP_563906.1 | 3 - 4 | 1007 | 70 | |
| NM_179320.2 | NP_849651.1 | 3 - 4 | 1055 | 70 | ||
| GenBank mRNA | AY034961.1 | - | 3 - 4 | 1007 | 70 | |
| AY113909.1 | - | 3 - 4 | 898 | 70 | ||
More Information
Set Membership: |
|

PHR1
Genomic Map