Assay Details
| Gene Symbol: PHR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332032.1 | NP_001321694.1 | 7 - 8 | 1253 | 101 | |
| NM_101109.1 | NP_563906.1 | 7 - 8 | 1325 | 101 | ||
| NM_179320.2 | NP_849651.1 | 7 - 8 | 1391 | 101 | ||
| GenBank mRNA | AY034961.1 | - | 7 - 8 | 1325 | 101 | |
| AY113909.1 | - | 7 - 8 | 1216 | 101 | ||
More Information
Set Membership: |
|

PHR1
Genomic Map