Assay Details
| Gene Symbol: THH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332124.1 | NP_001319003.1 | 9 - 10 | 1045 | 89 | |
| NM_179330.4 | NP_849661.1 | 10 - 11 | 917 | 89 | ||
| GenBank mRNA | AB057678.1 | - | 10 - 11 | 908 | 89 | |
| BT020325.1 | - | 10 - 11 | 799 | 89 | ||
| BT021134.1 | - | 10 - 11 | 788 | 89 | ||
More Information
Set Membership: |
|

THH1
Genomic Map