Assay Details
| Gene Symbol: ACA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332786.1 | NP_001321521.1 | 3 - 4 | 2571 | 102 | |
| NM_001332787.1 | NP_001321522.1 | 5 - 6 | 2510 | 102 | ||
| NM_102542.1 | NP_564295.1 | 5 - 6 | 2270 | 102 | ||
| NM_179385.2 | NP_849716.1 | 4 - 5 | 2541 | 102 | ||
More Information
Set Membership: |
|

ACA1
Genomic Map