Assay Details
| Gene Symbol: ACA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332786.1 | NP_001321521.1 | 5 - 6 | 3045 | 154 | |
| NM_001332787.1 | NP_001321522.1 | 7 - 8 | 2984 | 154 | ||
| NM_102542.1 | NP_564295.1 | 7 - 8 | 2744 | 154 | ||
| NM_179385.2 | NP_849716.1 | 6 - 7 | 3015 | 154 | ||
More Information
Set Membership: |
|

ACA1
Genomic Map