Assay Details
| Gene Symbol: AT1G40087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179426.4 | NP_849757.4 | 1 - 2 | 121 | 109 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G40087| Gene Symbol: AT1G40087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179426.4 | NP_849757.4 | 1 - 2 | 121 | 109 | |
Set Membership: | 
		   
					
						
							 |