Assay Details
| Gene Symbol: AT1G40087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179426.4 | NP_849757.4 | 5 - 6 | 765 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G40087| Gene Symbol: AT1G40087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179426.4 | NP_849757.4 | 5 - 6 | 765 | 102 | |
Set Membership: |
|