Assay Details
| Gene Symbol: AT1G43667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179438.2 | NP_849769.1 | 1 | 252 | 121 | |
| GenBank mRNA | AK118504.1 | - | 1 | 234 | 121 | |
| BT004689.1 | - | 1 | 153 | 121 | ||
More Information
Set Membership: |
|

AT1G43667
Genomic Map