Assay Details
| Gene Symbol: GCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198308.1 | NP_001185237.1 | 9 - 10 | 947 | 81 | |
| NM_001333703.1 | NP_001322789.1 | 9 - 10 | 994 | 81 | ||
| NM_001333704.1 | NP_001322788.1 | 9 - 10 | 987 | 81 | ||
| NM_179478.2 | NP_849809.2 | 10 - 11 | 980 | 81 | ||
More Information
Set Membership: |
|

GCT
Genomic Map