Assay Details
| Gene Symbol: CAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179491.3 | NP_849822.1 | 10 - 11 | 1821 | 78 | |
| GenBank mRNA | AF367304.1 | - | 10 - 11 | 1726 | 78 | |
| BT046174.1 | - | 10 - 11 | 1457 | 78 | ||
More Information
Set Membership: |
|

CAT2
Genomic Map