Assay Details
| Gene Symbol: CAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179491.3 | NP_849822.1 | 6 - 7 | 1159 | 86 | |
| GenBank mRNA | AF367304.1 | - | 6 - 7 | 1064 | 86 | |
| BT046174.1 | - | 6 - 7 | 795 | 86 | ||
More Information
Set Membership: |
|

CAT2
Genomic Map