Assay Details
| Gene Symbol: FLA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112101.1 | NP_187872.1 | 1 | 713 | 85 | |
| GenBank mRNA | DQ446657.1 | - | 1 | 713 | 85 | |
| Gene Symbol: emb2742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112102.3 | NP_187873.1 | 22 | 2493 | 85 | |
More Information
Set Membership: |
|

FLA14
Genomic Map