Assay Details
| Gene Symbol: emb2742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112102.3 | NP_187873.1 | 19 - 20 | 1577 | 77 | |
| GenBank mRNA | AY093110.1 | - | 19 - 20 | 1488 | 77 | |
| BT008360.1 | - | 18 - 19 | 1439 | 77 | ||
More Information
Set Membership: |
|

emb2742
Genomic Map