Assay Details
| Gene Symbol: emb2742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112102.3 | NP_187873.1 | 8 - 9 | 717 | 62 | |
| GenBank mRNA | AY093110.1 | - | 8 - 9 | 628 | 62 | |
| BT008360.1 | - | 7 - 8 | 579 | 62 | ||
More Information
Set Membership: |
|

emb2742
Genomic Map