Assay Details
| Gene Symbol: AT3G12710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112107.3 | NP_566433.1 | 2 - 3 | 702 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G12710| Gene Symbol: AT3G12710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112107.3 | NP_566433.1 | 2 - 3 | 702 | 86 | |
Set Membership: |
|