Assay Details
| Gene Symbol: PIE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338008.1 | NP_001326839.1 | 11 - 12 | 1688 | 81 | |
| NM_112117.5 | NP_187887.3 | 11 - 12 | 1700 | 81 | ||
| GenBank mRNA | AY279398.1 | - | 11 - 12 | 1163 | 81 | |
More Information
Set Membership: |
|

PIE1
Genomic Map