Assay Details
| Gene Symbol: PIE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338008.1 | NP_001326839.1 | 18 - 19 | 5857 | 91 | |
| NM_112117.5 | NP_187887.3 | 18 - 19 | 5869 | 91 | ||
| GenBank mRNA | AY279398.1 | - | 18 - 19 | 5332 | 91 | |
More Information
Set Membership: |
|

PIE1
Genomic Map