Assay Details
| Gene Symbol: PIE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338008.1 | NP_001326839.1 | 7 - 8 | 1242 | 132 | |
| NM_112117.5 | NP_187887.3 | 7 - 8 | 1254 | 132 | ||
| GenBank mRNA | AY279398.1 | - | 7 - 8 | 717 | 132 | |
More Information
Set Membership: |
|

PIE1
Genomic Map