Assay Details
| Gene Symbol: AT3G12860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112122.4 | NP_187892.2 | 6 - 7 | 1281 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G12860| Gene Symbol: AT3G12860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112122.4 | NP_187892.2 | 6 - 7 | 1281 | 78 | |
Set Membership: |
|