Assay Details
| Gene Symbol: AT3G12900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112126.2 | NP_187896.1 | 1 - 2 | 380 | 55 | |
| GenBank mRNA | DQ446658.1 | - | 1 - 2 | 267 | 55 | |
More Information
Set Membership: |
|

AT3G12900
Genomic Map