Assay Details
| Gene Symbol: SRF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112145.4 | NP_566444.4 | 11 - 12 | 1719 | 76 | |
| GenBank mRNA | AY518289.1 | - | 11 - 12 | 1719 | 76 | |
| Gene Symbol: AT3G13062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FJ154118.1 | - | 17 - 18 | 3254 | 76 | |
More Information
Set Membership: |
|

SRF4
Genomic Map