Assay Details
| Gene Symbol: SRF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112145.4 | NP_566444.4 | 13 - 14 | 1987 | 69 | |
| GenBank mRNA | AY518289.1 | - | 13 - 14 | 1987 | 69 | |
| Gene Symbol: AT3G13062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FJ154118.1 | - | 19 - 20 | 3522 | 69 | |
More Information
Set Membership: |
|

SRF4
Genomic Map