Assay Details
| Gene Symbol: SRF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112145.4 | NP_566444.4 | 4 - 5 | 507 | 67 | |
| GenBank mRNA | AY518289.1 | - | 4 - 5 | 507 | 67 | |
| Gene Symbol: AT3G13062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FJ154118.1 | - | 10 - 11 | 2042 | 67 | |
More Information
Set Membership: |
|

SRF4
Genomic Map